Learn More
Andwin Scientific NOVABLUE SINGLES COMP
SDP
Catalog No. 5089990317
Encompass_Preferred
NovaBlue Singles Competent Cells 11 reaction kit. Includes Test Plasmid and SOC Medium. For general cloning purposes. Has extremely high transformation efficiency blue/white screening of recombinants by lacZ alpha-complementation endA and recA mutations a
Catalog No. 50-899-90317 Supplier Andwin Scientific Supplier No. 701813
NovaBlue Singles Competent Cells 11 reaction kit. Includes Test Plasmid and SOC Medium. For general cloning purposes. Has extremely high transformation efficiency blue/white screening of recombinants by lacZ alpha-complementation endA and recA mutations a