Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NovaBright™ Phospha-Light™ EXP Assay Kit for SEAP (Secreted Placental Alkaline Phosphatase) Reporter Gene Detection

Catalog No. p-7078179
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 well plates, 960 assays
2 x 96 well plates, 192 assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10577 2 x 96 well plates, 192 assays
N10578 10 x 96 well plates, 960 assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10577 Supplier Invitrogen™ Supplier No. N10577
Only null left
Add to Cart
Add to Cart

Includes

Component solution containing CSPD substrate and chemiluminescence enhancer, placental alkaline phosphatase positive control

Chemiluminescent reporter gene assay for the ultrasensitive detection of secreted placental alkaline phosphatase or the non-secreted placental isozyme PLAP.

Sensitive detection of alkaline phosphatase reporter enzyme enables a large number of applications in many areas of life science research, including gene expression, viral function assays, vaccine development, development of viral vectors and gene delivery methods for gene therapy, in vivo gene expression monitoring and novel cellular functional assays. This SEAP assay is one of the easiest and fastest methods for optimizing transfection efficiency. This assay has been used for a wide variety of viral functional assays, including viral gene expression assays, viral replication, viral fusogenicity, virus neutralization and viral-mediated cell-cell fusion, and viral infectivity. Use of the SEAP reporter protein is very enabling for in vivo reporter gene assays, by assaying serum samples from transgenic, transfected or viral vector-infected animals. The NovaBright reporter gene assay system has been used to measure SEAP levels in sera from transgenic or transfected whole animals, including mouse, rat, marmoset, monkey and pig sera, and in chicken egg allantoic fluid. The mouse SEAP protein (mSEAP) has recently been developed for improved SEAP protein stability in transgenic mice, and the NovaBright SEAP assay has been used for sensitive detection of mSEAP . In addition to reporter gene (gene expression) applications, the assay system is used to measure SEAP as a functional reporter for receptor-ligand binding assays with a SEAP-ligand chimera, protease-mediated secretion, and for secretion pathway activity. The NovaBright assay system can be used for the cellular measurement of non-placental alkaline phosphatase as a biomarker.

This kit provides a much simpler procedure than other assays. The assay is homogenous and requires only two solutions both of which are supplied ready-to-use. Prolonged glow light emission kinetics provides a stable light signal for read-time flexibility. The assay signal is measured in a simple luminometer without injectors. SEAP is a reporter protein that is secreted into the cell culture medium and detected by testing aliquots of the medium. Since SEAP is released into the medium, cell lysis is unnecessary. However, the assay also allows the detection placental alkaline phosphase (PLAP) isoenzyme following cell lysis.

  • Second generation chemiluminescent reporter gene assay
  • Simple, fast protocol, high throughput-compatible
  • Ultrasensitive detection of secreted placental alkaline phosphatase
  • Compatibility with automated platforms with trigger solution/positions already in place
Specifications
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate)
  • Format: 96-well plate
  • High Throughput Compatibility: High Throughput-Compatible
  • Readout: End Point
  • Substrate: CSPD
  • Substrate Properties: Chemical Substrate
  • Substrate Type: AP (Alkaline Phosphatase) Substrate
  • Target Enzyme: Placental Alkaline Phosphatase
TRUSTED_SUSTAINABILITY

Specifications

Product Type Phosphatase Assay
Detection Method Enhanced Chemiluminescence
For Use With (Equipment) Luminometer (Microplate)
Content And Storage Component solution containing CSPD™ substrate and chemiluminescence enhancer (Store at 2°C to 6°C), placental alkaline phosphatase positive control (Store at ≤ -20°C)
High-throughput Compatibility High-throughput Compatible
Substrate CSPD™
Substrate Type AP (Alkaline Phosphatase) Substrate
Substrate Properties Chemical Substrate
Target Enzyme Placental Alkaline Phosphatase
Format 96-well Plate
Product Line NovaBright
Quantity 2 x 96 well plates, 192 assays
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.