Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ NovaClean™ Lab and Glass Cleaner
BIOPHARMAGREENER_CHOICESDP

Catalog No. 190353828 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Volume (English):
1 qt.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-035-3828 1 gal.
19-045-264 1 qt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-035-3828 Supplier Micronova™ Supplier No. NC2G
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready to use, low-ionic detergent can be applied directly to a variety of cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains.

Lab and glass cleaner is a low-ionic detergent, designed for use on cleanroom work-stations, laminar flow benches, acrylic and painted surfaces.

  • Ready to use, cleanroom surfactant
  • Filtered to 0.1 microns
  • Low mobile ions
  • Low non-volatile residue levels
  • Can be applied directly to a range of surfaces
  • Works well with glass, stainless steel, epoxy, acrylic, and vinyl curtains
  • Ideal for conductive flooring
  • Available in a 1 quart spray bottle and a 1 gallon refill bottle
  • Biodegradable
  • Free-rinsing

Specifications

Product Type Filtered Low Ionic Surfactant (0.1 μ)
Color-coded No
Form Liquid
Handle Type None
pH 7.1
Product Line NovaClean
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Boiling Point 99°C (210.2°F)
Care Instructions Avoid breathing vapor or mist and provide adequete ventilation when using.
Certifications/Compliance ISO 9001 Certified Facility; Certificate of Analysis
Color Clear
Container Type 1 Gallon Bottles
Content And Storage Store at ambient temperature. Avoid direct sunlight. Store in original container and keep tightly closed and seaqled until ready for use.
Density 1.025 g/mL at 25°C
Dimensions (L x W x H) 6 x 6 x 12 in.
Disposable Reusable
Filtered Filtered
Flammable No
For Use With (Application) Ready to use filtered cleanroom detergent, no mobile ions, free-rinsing for cleaning glass, stainless steel, acrylic and painted surfaces.
For Use With (Equipment) Mops and Buckets or Wipes and Sponge
Height (English) 12 in.
Height (Metric) 30.48 cm
Length (English) 6 in.
Length (Metric) 15.24 cm
Melting Point 0°C (32°F)
Packaging 4 bottles per poly-lined case
Quantity 4/Pk.
Resistant To The product is chemically stable. Under normal conditions of storage and use, hazardous reactions will not occur.
Size 13 x 13 x 13 in.
Vapor Pressure 2.3 kPa (17.1 mmHg)
Volume (English) 1 gal.
Volume (Metric) 3.79 L
Width (English) 6 in.
Width (Metric) 15.24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.