Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ 100mM IPTG Solution

Catalog No. 705273 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-527-3 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-527-3 Supplier MilliporeSigma™ Supplier No. 705273
Add to Cart
Add to Cart

Convenient, ready-to-use solution

  • The lac operon inducer IPTG (isopropyl-β-D- 1-thiogalactopyranoside) is recommended for blue/white screening by lacZ α- complementation with appropriate vectors and host strains, such as pSTBlue-1/NovaBlue, and for inducing protein expression with the pET System and other lac promoter-derived expression systems
  • The preparation is dioxane-free and supplied as a sterile aqueous solution, in convenient volumes (10 × 1.5mL)
TRUSTED_SUSTAINABILITY

Specifications

CAS 367-93-1
Content And Storage Store at −20°C
Quantity 15 mL
Synonym IPTG Solution, 100 mM
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.