Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ BacMagic™, Transfection Kit

Catalog No. 715463 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 Reactions/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-546-3 5 Reactions/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-546-3 Supplier MilliporeSigma™ Supplier No. 715463
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Creates baculovirus recombinants for recombinant protein expression in insect cells.

  • Provides positive selection for creating baculovirus recombinants in preparation for recombinant protein expression in insect cells
  • Eliminates tedious, time-consuming steps of plaque purification
  • AcNPV genome with portion of essential open reading frame (ORF) 1629 deleted
  • Prevents nonrecombinant virus from replicating in insect cells
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Multiple storage requirements
No. of Reactions 5 Reactions/Pk.
Quantity 5 Reactions/Pk.
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.