Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Bacteriophage CE6

Catalog No. 693903 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-390-3 0.2 mL
69-390-4 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-390-3 Supplier MilliporeSigma™ Supplier No. 693903
Add to Cart
Add to Cart

Contains T7 RNA polymerase gene.

EMD Millipore Novagen™ Bacteriophage CE6 is recombinant lambda phage.

  • Recombinant lambda phage
  • Provides source of T7 RNA polymerase to susceptible host cells carrying T7 expression plasmids
  • Has λcI857 immunity Sam7 mutation, which requires supF hosts for lytic growth
  • Provided as high titer lysate includes host strains LE392 (rK12- mK12+) ED8739 (rK12- mK12-) as glycerol stocks
  • 10mL size can induce up to 100mL culture
TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.2 mL
Storage Requirements ≤−70°C
Product Type Recombinant Lambda Phage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.