Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ BacVector™ 3000 Transfection Kit

Catalog No. 700773 Shop All MilliporeSigma Products
Change view
Click to view available options
BacVector™, 3000 Transfection Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
70-077-3 BacVector™, 3000 Transfection Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-077-3 Supplier MilliporeSigma™ Supplier No. 700773

missing translation for 'validMainframeMsg'

Designed for efficient, reliable construction of recombinant baculovirus.

  • Complete set of reagents kits designed for expression of target proteins in insect cells
  • CAS: Dimethyl Sulfoxide: 67-68-5
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Multiple Storage Conditions
No. of Reactions 12 Reactions/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.