Learn More
MilliporeSigma™ Novagen™ BetaFluor™ β-Galactosidase Assay Kit
Catalog No. 709793 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
70-979-3 1 Kit
1 options
Catalog No. 70-979-3 Supplier MilliporeSigma™ Supplier No. 709793
Add to Cart
Add to Cart
Designed for rapid, sensitive measurement of β-galactosidase activity in cell extracts.
EMD Millipore Novagen™ BetaFluor™ β-Galactosidase Assay Kit enables efficient passive extraction of Cell protein while maintaining optimal enzyme activity.
  • Configured for 96-well format
  • Highly amenable to high-throughput reporter gene expression analysis
  • Designed for fluorometric detection with 500fg sensitivity
TRUSTED_SUSTAINABILITY
Specifications
Kit Contents 1.6 mL 1 M DTT, 100 mL reaction buffer, 25 mL Reportasol™, 50 mL stop buffer, BetaRed substrate, Extraction buffer
Quantity 1 Kit