Learn More
MilliporeSigma™ Novagen™ Bovine Genomic DNA
Catalog No. 692313 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
69-231-3 100 μg
1 options
Catalog No. 69-231-3 Supplier MilliporeSigma™ Supplier No. 692313
Add to Cart
Add to Cart
Purified preparations of high molecular weight genomic DNA from a variety of organisms.
EMD Millipore Novagen™ Bovine Genomic DNA is qualified for genomic analysis library construction.
  • No inhibition of restriction enzyme activity
  • DNA free of contaminating RNA protein
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C