Learn More
MilliporeSigma™ Novagen™ Cleavage Control Protein
Catalog No. 690693 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-069-3 10 μg
1 options
Catalog No. 69-069-3 Supplier MilliporeSigma™ Supplier No. 690693
Add to Cart
Add to Cart
Monitors performance of thrombin or enterokinase cleavage conditions.
  • Cleaved fragments of thrombin enterokinase are easily visualized by SDS-PAGE
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Type Cleavage Control Protein
Quantity 10 μg