Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Clonables™ 2X Ligation Premix

Catalog No. p-7078292 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
2.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-573-3 50 μL
70-573-4 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-573-3 Supplier MilliporeSigma™ Supplier No. 705733
Only null left
Add to Cart
Add to Cart

Provides efficient ligation of any type of compatible DNA ends.

EMD Millipore Novagen™ Clonables™ 2X Ligation Premix is single solution containing optimized concentrations of T4 DNA ligase, buffer, stabilizer cofactors.

  • Tested for ligation of compatible 2 to 4 base cohesive DNA ends, blunt ends single-base overhangs found on some PCR products
  • Ligation is achieved in only 15 minutes
  • Compatible with transformation of any type of chemically competent cells
  • High performance, minimal pipetting requirement single-addition format suitable for high-throughput applications
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -70°C
Quantity 50 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.