Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Clonables™, Ligation/Transformation Kit

Catalog No. 705263 Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Reactions:
11 Reactions/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
70-526-3 11 Reactions/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-526-3 Supplier MilliporeSigma™ Supplier No. 705263
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Supports ligation of any type of DNA cohesive or blunt ends.

EMD Millipore Novagen™ Clonables™ Ligation/Transformation Kit enables convenient, dependable, high-efficiency ligation transformation of any compatible DNA ends.

  • Fast 15 minute ligation, eight minute transformation with ampicillin selection
  • Ligation components premixed to decrease pipetting steps increase reliability
  • One reaction, optimized for cohesive ends, single base overhangs blunt ends
  • Single-use competent cells eliminate need to aliquot, freeze/thaw, or waste partially used vials
  • Compatible with PCR buffer, TE, restriction enzyme buffer or End conversion mix
  • Features unique, universal Ligation Premix, containing ligase, buffer, cofactors
  • Ligated DNA is transformed into NovaBlue Singles™ competent Cells can be used with a variety of cloning vectors
  • CAS: Dimethyl Sulfoxide: 67-68-5, Tris Base:77-86-1
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

No. of Reactions 11 Reactions/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.