Learn More
MilliporeSigma™ Novagen™ D-Desthiobiotin
Catalog No. 716103 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
71-610-3 1 g
1 options
Catalog No. 71-610-3 Supplier MilliporeSigma™ Supplier No. 716103
Add to Cart
Add to Cart
Provides elution of Strep·Tag™ II proteins from biotin-binding site of Strep·Tactin™ resins
  • Lyophilized Powder
  • For preparation of elution buffer
  • Provides gentle elution of streptovidin tagged II proteins from biotin-binding site of resins
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 g