Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ DNA Ligation Kit

Catalog No. 698383 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-838-3 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-838-3 Supplier MilliporeSigma™ Supplier No. 698383
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tested for PCR cloning, ligation of cDNA, PCR products into plasmid phage lambda vectors.

EMD Millipore Novagen™ DNA Ligation Kit provides convenient, reproducible ligation of DNA fragments.

  • Provides necessary components for convenient, reproducible ligation of DNA fragments
  • Contains components for 50 blunt-end or single-base overhang ligations, or up to 500, 2 to 4 base cohesive end ligations
  • CAS: Dithiothreitol: 3483-12-3
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.