Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Ekapture™ Agarose

Catalog No. 690684 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1.5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-068-4 10 mL
69-068-3 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-068-4 Supplier MilliporeSigma™ Supplier No. 690684
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Intended for rapid affinity-based capture.

EMD Millipore Novagen™ Ekapture™ Agarose is used for removal of enterokinase.

  • Removed with >99% efficiency from reaction by affinity capture
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Type Agarose
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.