Learn More
MilliporeSigma™ Novagen™ Enterokinase Cleavage Capture Kit
Catalog No. 690673 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
69-067-3 1 Kit
1 options
Catalog No. 69-067-3 Supplier MilliporeSigma™ Supplier No. 690673
Add to Cart
Add to Cart
Designed for specific cleavage of fusion proteins.
EMD Millipore Novagen™ Enterokinase Cleavage Capture Kit follows rapid, affinity-based capture for removal of enterokinase.
  • Features amino terminal S Tag™ sequence
  • Enables sensitive detection of 16 kd proteolytic product with Western Blot reagents
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Multiple storage requirements
Type Enterokinase Cleavage Capture Kit
Quantity 1 Kit