Learn More
MilliporeSigma™ Novagen™ Factor Xa Cleavage Capture Kit
GSA_VA
Catalog No. 690373 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
69-037-3 1 Kit
1 options
Catalog No. 69-037-3 Supplier MilliporeSigma™ Supplier No. 690373
Add to Cart
Add to Cart
Includes
100μg Restriction Grade Factor Xa, 2mL Factor Xa Dilution/Storage buffer, 5mL 10X Factor Xa Cleavage/Capture buffer, 2 x 25mL Xarrest™ Agarose, 10μg Xa Cleavage Control Protein, 10/pk. Spin Filters, 2mL capacity
Designed for specific cleavage of fusion proteins.
EMD Millipore Novagen™ Factor Xa Cleavage Capture Kit follows convenient affinity-based capture.
  • Convenient affinity-based capture removal of Factor Xa
  • Features an amino terminal S·Tag™ sequence enabling sensitive detection of the 17 kd proteolytic product with Western Blot reagents
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Type Factor Xa Cleavage Capture Kit
Quantity 1 Kit