Learn More
MilliporeSigma™ Novagen™ FastPlax™ Titer Kit
Catalog No. 708503 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
70-850-3 1 Kit
1 options
Catalog No. 70-850-3 Supplier MilliporeSigma™ Supplier No. 708503
Add to Cart
Add to Cart
Includes
  • 2 x 50mL 10X TBST (100mM Tris-HCl, pH 8.0, 1.5M NaCl, 1% Tween 20)
  • 6mL 10% Gelatin (contains 0.02% azide)
  • 10μL FastPlax Antibody
  • 300μL Goat-Anti-Mouse β-Galactosidase Conjugate
  • 300μl NBT (83mg/mL in
Designed for determination of baculovirus titers.
EMD Millipore Novagen™ FastPlax™ Titer Kit detection is based on high affinity monoclonal antibody against gp64, which is added directly to fixed cells.
  • Determination of baculovirus titers in 24 to 48 hours
  • Detected with goat Anti-Mouse IgG β-Galactosidase conjugate followed by enhanced color Development with X-Gal/NBT substrates
  • Plaques are clearly distinguished by their dark blue color
  • Used to detect appearance of AcNPV gp64 glycoprotein on surface of infected cells in 8 to 24 hrs post-infection
  • Packaging: Plastic Ampoule
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit