Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ GeneJuice™, Transfection Reagent

Catalog No. 709676 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.3 mL
1 mL
5 mL
10 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-967-6 5 mL
70-967-3 1 mL
70-967-4 10 mL
70-967-5 0.3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 70-967-6 Supplier MilliporeSigma™ Supplier No. 709676
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enables efficient DNA transfer in both stable transient transfections of eukaryotic cells.

EMD Millipore Novagen™ GeneJuice™, Transfection Reagent is non-lipid based chemical transfection reagent.

  • Efficient DNA transfer for both stable transient transfections
  • Minimal cellular toxicity
  • Compatibility with both serum-containing serum-free media
  • Simple protocol
  • Suitable for high-throughput transfection in multi-well plate format
  • CAS: Ethanol: 64-17-5
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.