Learn More
Description
- Protein purification kit
- CAS: Nickel(II) Sulfate Hexahydrate: 10101-97-0, Ethylenediaminetetraacetic Acid: 60-00-4
- Packaging: Glass Bottle
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Format | Kit |
| Product Type | Protein Purification Kit |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.