Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Human Male Genomic DNA

Catalog No. 705723 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-572-3 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-572-3 Supplier MilliporeSigma™ Supplier No. 705723
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For genomic analysis, including PCR and library construction. It is free of contaminating RNA and protein.

  • High molecular weight
  • High purity DNA for any application
  • Isolation protocol minimizes shearing and removes contaminants that inhibit enzymatic procedures
  • Pipet with a wide-bore pipet tip to minimize shearing
  • At least 90% of DNA is greater than 100kbp in size, as analyzed by CHEF gel electrophoresis (>50kbp for S. cerevisiae)
  • No inhibition of restriction enzyme activity
TRUSTED_SUSTAINABILITY

Specifications

pH 8
Product Type Human Male Genomic DNA
Reagents 10mM Tris-HCl, 1mM EDTA (TE)
Source Multiple organisms
Content And Storage 2°C to 8°C
Quantity 100 μg
For Use With (Application) PCR, Library construction

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.