Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Insect PopCulture™ Reagent

Catalog No. 711874 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-187-4 250 mL
71-187-3 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-187-4 Supplier MilliporeSigma™ Supplier No. 711874
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Extracts protein directly from insect cell cultures.

EMD Millipore Novagen™ Insect PopCulture™ Reagent is detergent-based lysis reagent without the need for centrifugation.

  • Recovers both protein released into medium and intracellular protein
  • Increases processing efficiency and target protein yields
  • Amenable for automated expression-level screening
  • Compatible with Ni-NTA His·Bind™ affinity purification method
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to -8°C
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.