Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Magnetight™ Oligo(dT) Particles

Catalog No. 695933 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
8 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-593-3 8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-593-3 Supplier MilliporeSigma™ Supplier No. 695933
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains covalently attached oligo(dT)25 to directly bind mRNA.

EMD Millipore Novagen™ Magnetight Oligo(dT) Particles are encapsulated paramagnetic particles.

  • Treated to minimize non-specific interactions enable quantitative recovery of intact mRNA
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Magnetight Oligo(dT) Particles
Packaging Glass Bottle
Quantity 8 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.