Learn More
MilliporeSigma™ Novagen™ Mouse Ig-Primer Set
GSA_VA
Catalog No. 698313 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Catalog No. Quantity
69-831-3 1 Set
1 options
Catalog No. 69-831-3 Supplier MilliporeSigma™ Supplier No. 698313
Add to Cart
Add to Cart
Designed for amplification of immunoglobulin variable region cDNAs from mouse sources.
EMD Millipore Novagen™ Mouse Ig-Primer Set is used in PCR to amplify variable regions of light heavy chain cDNAs.
  • Amplification of immunoglobulin variable regions
  • Powerful method enables rapid, precise analysis of hybridomas, myeloma cell progression heterogeneity, idiotype networks, B-cell clonal ontogeny, phylogenetic relationships
  • Packaging: Glass Bottle
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Quantity 1 Set