Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ NoShift™ Transcription Factor Assay Kit

Catalog No. 713773 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-377-3 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-377-3 Supplier MilliporeSigma™ Supplier No. 713773

missing translation for 'validMainframeMsg'

Detects DNA binding protein in microassay plate based method.

EMD Millipore Novagen™ NoShift™ Transcription Factor Assay Kit is used with oligonucleotides defining protein binding consensus sequence.

  • Analyzes interactions between proteins DNA
  • Versatile, enables simultaneous analysis of multiple binding factors in less than five hours without use of radioisotopes
  • Bound transcription factor is detected with specific antibody followed by second antibody HRP conjugate chromogenic substrate
  • Applications:
    • Designed for use with oligonucleotides defining protein binding consensus sequence
    • CAS: Dithiothreitol: 3483-12-3, Tetramethylbenzidine: 54827-17-7
    • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Multiple storage requirements
No. of Reactions 96 Reactions/Pk.
Quantity 1 Kit
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.