Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ NovaQUANT™ Human Mitochondrial to Nuclear DNA Ratio Kit

Catalog No. 726201KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
726201KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 726201KIT Supplier MilliporeSigma™ Supplier No. 726201KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compares levels of nuclear to mitochondrial DNA (mtDNA) in human DNA sample. It has optimized qPCR protocol for sensitive, specific and linear detection of mitochondrial or nuclear target genes.

  • Eliminate detection of unwarranted mitochondrial pseudogenes in your data
  • Accurately determine DNA ratios based on matched primer pair amplification efficiencies
  • Pre-aliquoted primers for automating your protocols
  • Contains two qPCR plates with pre-aliquoted PCR primer pairs, Human wt DNA, and Human rho zero DNA
  • Contains enough DNA for 48 tests (using two 96-well microplates)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Human Mitochondrial to Nuclear DNA Ratio Kit
Content And Storage All materials in this kit can be kept at 4°C for up to three months; Control DNAs at -20°C for long term storage.
Quantity 1 Kit

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.