Learn More
MilliporeSigma™ Novagen™ NucBuster™ Protein Extraction Kit
Catalog No. 711833 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
71-183-3 1 Kit
1 options
Catalog No. 71-183-3 Supplier MilliporeSigma™ Supplier No. 711833
Add to Cart
Add to Cart
Prepares nuclear extracts from mammalian cells.
EMD Millipore Novagen™ NucBuster™ Protein Extraction Kit is based on two proprietary detergent-based solutions.
  • Ready to use
  • Compatible with electrophoretic mobility shift assays (EMSA)
  • Optimized for cell lysis removal of cytoplasmic components
  • Optimized for extraction of nuclear proteins
  • Free of stickiness associated with release of genomic DNA
  • No Dounce homogenization, dialysis required
  • Single microcentrifuge tube
  • Scalable protocol
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Content And Storage 2° to −8°C