Learn More
MilliporeSigma™ Novagen™ Overnight Express™ Autoinduction NMR Medium-15N
Catalog No. 717594 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L Kit
5 L Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-759-4 5 L Kit
71-759-3 1 L Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-759-4 Supplier MilliporeSigma™ Supplier No. 717594
Add to Cart
Add to Cart
Provides reagents to make a chemically defined autoinducing liquid culture medium used to produce [U-15N] labeled proteins for NMR spectroscopy. Novagen™ Overnight Express™ Autoinduction NMR Medium- 15N system includes four concentrated sterile solutions.
  • Autoinduction method is based on the differential metabolism of carbon sources to promote growth to high cell density
  • Automatically induce protein expression under control of lac promoters, eliminating the need to monitor cell densities or add inducer (IPTG)
  • Includes four concentrated sterile solutions:
  • OnEx NMR Solution 1 is a blend of carbon sources optimized for tightly regulated uninduced growth to high density, followed by autoinduction
  • OnEx NMR Solution 2 is a buffer and nitrogen blend that mediates metabolic acid production and provides additional nitrogen for increased protein synthesis
  • OnEx Solution 3 provides magnesium for maximal cell density
  • OnEx Solution 4 provides trace metals to minimize growth limitations associated with mineral deficiencies and satisfy the requirements of metal-containing target proteins, even at high expression levels
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 L Kit
Content And Storage Multiple storage requirements. Ok to freeze.
Product Type NMR Medium-15N