Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Overnight Express™ Autoinduction NMR Medium - 15N 13C

Catalog No. 717893 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-789-3 1 L Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-789-3 Supplier MilliporeSigma™ Supplier No. 717893
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to optimize cell growth and protein expression before preparing isotopically labeled proteins for NMR spectroscopy. MilliporeSigma™; Novagen™ Overnight Express™ Autoinduction NMR Optimization Medium is based on the differential metabolism of carbon sources to promote growth to high cell density.

  • Automatically induce protein expression under control of lac promoters, eliminating the need to monitor cell densities or add inducer (IPTG)
  • Includes four concentrated sterile solutions:
  • OnEx Dual-NMR Solution 1 is a blend of carbon sources optimized for tightly regulated uninduced growth to high density, followed by autoinduction
  • OnEx NMR Solution 2 is a buffer and nitrogen blend that mediates metabolic acid production and provides additional nitrogen for increased protein synthesis
  • OnEx Solution 3 provides magnesium for maximal cell density
  • OnEx Solution 4 provides trace metals to minimize growth limitations associated with mineral deficiencies and satisfy the requirements of metal-containing target proteins, even at high expression levels
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 L Kit
Content And Storage Multiple storage requirements. Ok to freeze.
Product Type Autoinduction NMR Medium - 15N 13C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.