Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Overnight Express™ Autoinduction System 1
GSA_VA

Catalog No. 713003 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-300-3 1 L
71-300-4 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-300-3 Supplier MilliporeSigma™ Supplier No. 713003
Add to Cart
Add to Cart

Induces protein expression without monitoring cell density.

EMD Millipore Novagen™ Overnight Express™ Autoinduction System 1 is based on media components that are metabolized differentially to promote growth.

  • Blend of carbon sources optimized for tightly regulated uninduced growth to high cell density followed by high-level induction
  • Provides magnesium for maximal cell density
  • Convenient for routine expression of proteins in multiple cultures
  • Ideal for high-throughput parallel analysis of protein expression, solubility, purification from multiple expression clones
  • Allows rapid optimization of the host-vector combinations, expression conditions
  • Purification parameters for high-throughput production of proteins for structural or functional analysis
  • Packaging: Glass Bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 L
Content And Storage 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.