Learn More
MilliporeSigma™ Novagen™ PCR Markers, 50–2000 bp
Catalog No. 692783 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 Lanes
Catalog No. Quantity
69-278-3 50 Lanes
1 options
Catalog No. 69-278-3 Supplier MilliporeSigma™ Supplier No. 692783
Add to Cart
Add to Cart
Includes
Separate vial of 6X Loading Buffer
Used for characterizing small DNA products.
EMD Millipore Novagen™ PCR Markers, 50–2000 bp is mixture of eight defined DNA fragments ranging from 50 to 2000 bp.
  • Markers are available in convenient size intervals
  • Each vial is enough for 50 lanes on TBE or TAE agarose gels stained with ethidium bromide
  • 5μl produces bands of even intensity that are bright, sharp easy to photograph
TRUSTED_SUSTAINABILITY
Specifications
Description Locates fragments from 50 to 2000 bp
Quantity 50 Lanes
Storage Requirements 2° to 8°C
Product Type DNA Molecular Weight Marker