Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Pellet Paint™ NF Co-Precipitant
GSA_VA

Catalog No. 707484 Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Reactions:
125
1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
70-748-4 1000
70-748-3 125
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-748-4 Supplier MilliporeSigma™ Supplier No. 707484
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Useful substitute for the original Pellet Paint Co-Precipitant in applications where fluorescent detection is used

  • Efficient and rapid precipitation of BigDye™ cycle sequencing products
  • Efficient removal of dye terminators
  • Direct visualization and tracking of precipitated material
  • No effect on sequencing reaction
  • Substitute for original Pellet Paint™ Co-Precipitant for fluorescent detection applications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at −20°C
Label or Dye Non-fluorescent
No. of Reactions 1000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.