Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Perfect DNA™ Markers - 0.1-12 kbp

Catalog No. 700873 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Lanes/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-087-3 100 Lanes/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-087-3 Supplier MilliporeSigma™ Supplier No. 700873
Add to Cart
Add to Cart

Includes

Extra vial of 6X DNA Gel Loading Buffer

Used in plasmid cloning.

EMD Millipore Novagen™ Perfect DNA™ Markers - 0.1-12 kbp contain sets of DNA species.

  • Easy-to-remember sizes for agarose gel analysis
  • Uniform b intensities
  • Easily identifiable reference bs, which are useful for instant b identification
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Quantity 100 Lanes/Pk.
Storage Requirements −20°C
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.