Learn More
MilliporeSigma™ Novagen™ Perfect Protein™ Markers, 10-225 kd
Catalog No. 690793 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Lanes
Catalog No. Quantity
69-079-3 100 Lanes
1 options
Catalog No. 69-079-3 Supplier MilliporeSigma™ Supplier No. 690793
Add to Cart
Add to Cart
Designed for routine use in SDS-polyacrylamide gel electrophoresis, the Perfect Protein Markers enable highly accurate size determination of unknown samples.
EMD Millipore Novagen™ Perfect Protein Markers, 10-225 kd are set of recombinant proteins with defined sizes at convenient intervals.
  • Optimized for use with Coomassie blue staining
  • Adjusted amounts can also be used with silver staining, fluorescent dyes
  • Includes protein sizes of 15, 25, 35, 50, 75, 100, 150 kd
  • Each vial contains 400μg protein
  • Includes two additional proteins, 10 kd 225 kd, for applications requiring broader size range
  • Each vial contains 500μg protein
TRUSTED_SUSTAINABILITY
Specifications
Description Markers, 10-225 kd
Quantity 100 Lanes
Storage Requirements −20°C
Product Type Protein Molecular Weight Marker