Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Perfect Protein™, Western Markers

Catalog No. 699593 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Lanes/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-959-3 25 Lanes/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-959-3 Supplier MilliporeSigma™ Supplier No. 699593
Add to Cart
Add to Cart

Detects markers by adding S-protein or His·Tag™.

EMD Millipore Novagen™ Perfect Protein, Western Markers can be used with colorimetric chemiluminescent AP or HRP substrates.

  • S-protein conjugate does not interfere with antibodies or streptavidin detection
  • Recombinant, unmodified markers gives sharp, accurately sized bs
  • Markers are supplied at working dilution in gel loading buffer
  • Concentration optimized for Western detection
  • Protein sizes are 15, 25, 35, 50, 75, 100 150 kd
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Quantity 25 Lanes/Pk.
Storage Requirements −20°C
Product Type Western Blot Protein Molecular Weight Markers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.