Description
- No restriction enzymes or special primers
- Compatible with any DNA polymerase
- Independent of 3′-dA addition or sequence context of DNA ends
- Blue/white screening with seven different vectors, including pETBlue expression vectors
- Simple protocol takes less than one hour from PCR product to plating transformants
Specifications
Specifications
| Product Type | pETBlue-1 Cloning Kit |
| Content And Storage | ≤-70°C |
| Quantity | 20 Reactions |