Learn More
MilliporeSigma™ Novagen™ Perfectly Blunt™ pETBlue-2 Cloning Kit
Catalog No. 706363 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
Catalog No. Quantity
70-636-3 20 Reactions
1 options
Catalog No. 70-636-3 Supplier MilliporeSigma™ Supplier No. 706363
Add to Cart
Add to Cart
For simplified cloning of DNA generated by PCR using any type of DNA polymerase
  • No restriction enzymes or special primers
  • Compatible with any DNA polymerase
  • Independent of 3′-dA addition or sequence context of DNA ends
  • Blue/white screening with seven different vectors, including pETBlue expression vectors
  • Simple protocol takes less than one hour from PCR product to plating transformants
TRUSTED_SUSTAINABILITY
Specifications
Product Type pETBlue-2 Cloning Kit
Content And Storage ≤-70°C
Quantity 20 Reactions