Learn More
MilliporeSigma™ Novagen™ Perfectly Blunt™ pSTBlue-1 Giga Cloning Kit
Catalog No. 712293 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
Catalog No. Quantity
71-229-3 20 Reactions
1 options
Catalog No. 71-229-3 Supplier MilliporeSigma™ Supplier No. 712293
Add to Cart
Add to Cart
For increased transformation efficiency
  • No restriction enzymes or special primers
  • Compatible with any DNA polymerase
  • Independent of 3′-dA addition or sequence context of DNA ends
  • Blue/white screening with seven different vectors, including pETBlue expression vectors
  • Simple protocol takes less than one hour from PCR product to plating transformants
TRUSTED_SUSTAINABILITY
Specifications
Product Type pSTBlue-1 Giga Cloning Kit
Content And Storage ≤-70°C
Quantity 20 Reactions