Learn More
MilliporeSigma™ Novagen™ pET-32a(+) DNA
Catalog No. 690153 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-015-3 10 μg
1 options
Catalog No. 69-015-3 Supplier MilliporeSigma™ Supplier No. 690153
Add to Cart
Add to Cart
Designed for cloning high-level expression of peptide sequences.
EMD Millipore Novagen™ pET-32a(+) DNA has cloning sites available for producing fusion proteins.
  • Peptide sequences fused with 109aa Trx Tag™ thioredoxin protein
  • Cloning sites are available for producing fusion proteins containing cleavable His Tag™ S Tag™ sequences for detection purification
  • pET Vectors are supplied as purified plasmid DNA (10μg)
  • pET DNA also includes an Induction Control strain
  • Packed in Glass Bottle
TRUSTED_SUSTAINABILITY
Specifications
Type pET-32a(+) DNA
Quantity 10 μg
Content And Storage ≤-70°C