Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ PopCulture™ Reagent

Catalog No. 710924 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
15 mL
75 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-092-4 75 mL
71-092-3 15 mL
71-092-5 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71-092-4 Supplier MilliporeSigma™ Supplier No. 710924
Add to Cart
Add to Cart

Formulated to extract proteins from E. coli cells directly in culture medium.

EMD Millipore Novagen™ PopCulture™ Reagent is buffered mixture of concentrated detergents.

  • Perforates E. coli cell wall without denaturing soluble protein protects protein from pH extremes
  • Assayed directly or purified by adding affinity matrix
  • Performed in single tube without need for columns or centrifugation
  • Applications:
    • High throughput (HT) robotic processing of samples for proteomics research
    • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Quantity 75 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.