Learn More
MilliporeSigma™ Novagen™ ProteoExtract™ S-PEK Antibody Control Kit
Catalog No. 717713 Shop All MilliporeSigma Products
Change view
Click to view available options
ProteoExtract™ S-PEK Antibody Control Kit
Catalog No.
71-771-3 ProteoExtract™ S-PEK Antibody Control Kit
1 options
Catalog No. 71-771-3 Supplier MilliporeSigma™ Supplier No. 717713
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Used to recognize proteins specific for each of four sub-cellular fractions.
  • Set of four monoclonal antibodies that recognize proteins specific for each of four sub-cellular fractions
Applications:
  • Used in Western blot procedures to confirm proper sub-fractionation of cellular proteomes