Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ ProteoExtract™ Transmembrane Protein Extraction Kit

Catalog No. 717723 Shop All MilliporeSigma Products
Change view
Click to view available options
Sufficient For:
20 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
71-772-3 20 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-772-3 Supplier MilliporeSigma™ Supplier No. 717723

missing translation for 'validMainframeMsg'

Enables mild efficient extraction of transmembrane proteins from mammalian cells tissues.

  • Detergent free novel chemistry
  • Enriched in membrane proteins large protein complexes
  • Compatible with enzyme assays, non-denaturing SDS-based gel electrophoresis, Western blotting immunoprecipitation
  • Enriched fraction appropriate for mass spectroscopy based analysis after tryptic in-gel digest
TRUSTED_SUSTAINABILITY

Specifications

Sufficient For 20 Samples
Kit Contents Two different extraction reagents for protein-dependent optimization, protease inhibitor cocktail, and appropriate buffers
For Use With (Application) Extraction of transmembrane proteins from mammalian cells and tissues
Content And Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.