Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ ProteoJuice™ Protein Transfection Reagent

Catalog No. 712813 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-281-3 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-281-3 Supplier MilliporeSigma™ Supplier No. 712813
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delivers protein into mammalian cells.

EMD Millipore Novagen™ ProteoJuice™ Protein Transfection Reagent is effective reagent with minimal toxicity and broad cell specificity.

  • Forms non-covalent complex with protein
  • Protects against degradation by endosomal proteases
  • Ensures delivery of intact protein inside cell
  • Live cells can be examined without need for cell fixation

  • Applications:

    • Used for functional studies, delivering bioactive or inhibitory proteins, screening peptide libraries, studying subcellular location turnover, transient complementation of suppressed gene expression
    • Used to deliver large proteins (such as antibodies), small proteins (such as histones), peptides
    • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to -8°C
Quantity 0.125 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.