Learn More
MilliporeSigma™ Novagen™ pSTBlue-1 AccepTor™ Vector Kit
Catalog No. 705954 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-595-4 40 Reactions
70-595-3 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-595-4 Supplier MilliporeSigma™ Supplier No. 705954
Add to Cart
Add to Cart
Designed to simplify the PCR cloning process by using non-proofreading thermostable DNA polymerases, which leave single 3'-dA overhangs on the reaction products. MilliporeSigma™; Novagen™ pSTBlue-1 AccepTor™ Vector Kit are supplied in a ready-to-ligate format.
  • No restriction enzymes or special primers
  • Direct ligation of PCR product with vector
  • Compatible with polymerases that leave single 3´-dA overhangs
  • Blue/white screening with pSTBlue-1 or pETBlue™-1 vectors
  • Simple protocol takes as little as 40 minutes from PCR product to plating transformants
TRUSTED_SUSTAINABILITY
Specifications
Product Type pSTBlue-1 Vector Kit
Content And Storage Store at ≤70°C
Quantity 40 Reactions