Learn More
MilliporeSigma™ Novagen™ pTandem™-1 Vector
Catalog No. 712833 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20μg
Catalog No. Quantity
71-283-3 20μg
1 options
Catalog No. 71-283-3 Supplier MilliporeSigma™ Supplier No. 712833
Coexpression of two genes from one vector in mammalian cells from a bicistronic RNA. MilliporeSigma™; Novagen™ pTandem™-1 Vector contains separate multiple cloning sites located on both sides of an internal ribosome binding site (IRES).
  • Features provide for convenient insertion of two open reading frames and efficient translation of both encoded proteins from a single bicistronic mRNA
TRUSTED_SUSTAINABILITY
Specifications
Product Type 1 Vector
Content And Storage Store at -20°C. Ok to freeze.
Quantity 20μg