Learn More
MilliporeSigma™ Novagen™ pTriEx™-3 Neo DNA
Catalog No. 709313 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20μg
Catalog No. Quantity
70-9313 20μg
1 options
Catalog No. 70-9313 Supplier MilliporeSigma™ Supplier No. 709313
Add to Cart
Add to Cart
Designed to allow rapid characterization of target genes in E. coli, insect and vertebrate cells. MilliporeSigma™; Novagen™ pTriEx™-3 Neo DNA allows rapid selection of stable transfected vertebrate cells expressing high levels of target gene.
  • Transient vertebrate expression is mediated by CMV immediate early enhancer and promoter
  • For expression in insect cells, pTriEx-3 Neo contains flanking baculovirus sequences to permit generation of recombinant baculoviruses using the BacVector™ System
  • In baculovirus-infected insect cells, expression is driven by the very late p10 promoter
  • Expression in E. coli is regulated by tightly controlled T7lac promoter
  • pTriEx recombinant plasmids can be transferred into (DE3) pLacI host that allows IPTG based induction
TRUSTED_SUSTAINABILITY
Specifications
Product Type 3 Neo DNA
Content And Storage Store at -20°C. Ok to freeze.
Quantity 20μg