Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ pTriEx™-6 DNA

Catalog No. 715593 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-559-3 20μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-559-3 Supplier MilliporeSigma™ Supplier No. 715593
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to allow rapid characterization of target genes in multiple expression systems. MilliporeSigma™; Novagen™ pTriEx™-6 DNA can be used to test expression in E. coli, insect and mammalian cells.

  • Transient vertebrate expression is mediated by CMV immediate early enhancer and promoter
  • For expression in insect cells, pTriEx-6 contains flanking baculovirus sequences to permit generation of recombinant baculoviruses using the BacVector™ System
  • In baculovirus-infected insect cells, expression is driven by the very late p10 promoter
  • Expression in E. coli is regulated by tightly controlled T7lac promoter
  • pTriEx recombinant plasmids can be transferred into (DE3) pLacI host that allows IPTG based induction
TRUSTED_SUSTAINABILITY

Specifications

Format Glass bottle
Product Type 6 DNA
Content And Storage Store at -20°C. Ok to freeze.
Quantity 20μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.