Learn More
MilliporeSigma™ Novagen™ PureGenome™ Next Generation Sequencing Library Amplifier
Catalog No. PGN0031EA Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Each/Kit
Catalog No. Quantity
PGN0031EA 1 Each/Kit
1 options
Catalog No. PGN0031EA Supplier MilliporeSigma™ Supplier No. PGN0031EA
Optimized for NGS library preparation.
  • Validated for sequencing compatibility with Illumina™ instruments
  • Ultra high fidelity fast yielding KOD hot start DNA polymerase
  • Rapid amplification of NGS libraries for ensuring required inputs for NGS instrument sequencing high quality data output
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Quantity 1 Each/Kit