Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Recombinant Enterokinase

Catalog No. 690663 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-066-3 50 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-066-3 Supplier MilliporeSigma™ Supplier No. 690663
Add to Cart
Add to Cart

Purified preparation of catalytic subunit of bovine enterokinase.

EMD Millipore Novagen™ Recombinant Enterokinase recognizes identical cleavage site as native enzyme.

  • Produced in E. coli
  • Purified to yield the higher activity
  • Qualified for specific cleavage of appropriate fusion proteins
  • Supplied as solution in 50% glycerol
  • Unit of Definition One unit is defined as the amount of enzyme that will cleave 50μg of fusion protein in 16 hours at 23°C, in a buffer containing 20mM Tris-HCl pH 7.4, 50mM NaCl, 2mM CaCl2
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Type Recombinant Enterokinase
Quantity 50 U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.