Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ S-protein Agarose
GSA_VA

Catalog No. 697044 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-704-4 10 mL
69-704-3 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-704-4 Supplier MilliporeSigma™ Supplier No. 697044
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Retains specifically S·Tag™ fusion proteins.

EMD Millipore Novagen™ S-protein Agarose binds in both column or batch mode.

  • Elution is done under slightly denaturing conditions or using specific protease, such as thrombin or enterokinase, with appropriate recombinant proteins
  • Capacity varies is based on size folding characteristics of given target protein
  • Packaging: Glass Bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.